Learn More
Wilmad Labglass FLSK VOL CL A 100ML PLS STPR
Supplier:  Wilmad Labglass LG8116112
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 100mL +/- 0.08mL capacity. Stopper Size: 13.Calibrated to contain at 20degreeC.Flask is provided with plastic closed-bottom Stopper.
Catalog No. 50-802-972