Learn More
Wilmad Labglass FLSK VOL CL A 10ML PLS STPR
Supplier:  Wilmad Labglass LG8116106
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 10mL +/- 0.02mL capacity. Stopper Size: 9. Calibrated to contain at 20degreeC. Flask is provided with plastic closed-bottom Stopper..
Catalog No. 50-802-911
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product