Learn More
Wilmad Labglass FLSK VOL CL A 200ML ST13 STPR
Supplier:  Wilmad Labglass LG8110114
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 200mL +/- 0.10mL capacity. Stopper Size: 13.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.
Catalog No. 50-804-489