Learn More
Wilmad Labglass FLSK VOL CL A 250ML PLS STPR
Supplier:  Wilmad Labglass LG8116116
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 250mL +/- 0.12mL capacity. Stopper Size: 16.Calibrated to contain at 20degreeC.Flask is provided with plastic closed-bottom Stopper..
Catalog No. 50-807-803
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product