Learn More
Wilmad Labglass FLSK VOL CL A 25ML ST9 STPR
Supplier:  Wilmad Labglass LG8110108
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 25mL +/- 0.03mL capacity. Stopper Size: 9. Calibrated to contain at 20degreeC. Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.
Catalog No. 50-803-144