Learn More
Wilmad Labglass FLSK VOL CL A 3000ML PLS STPR
Supplier:  Wilmad Labglass LG8116124
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 3000mL +/- 1.0mL capacity. Stopper Size: 32.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.
Catalog No. 50-803-913