Learn More
Wilmad Labglass FLSK VOL CL A 500ML PLS STPR
Supplier:  Wilmad Labglass LG8116118
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 500mL +/- 0.20mL capacity. Stopper Size: 19.Calibrated to contain at 20degreeC.Flask is provided with plastic closed-bottom Stopper..
Catalog No. 50-803-376