Learn More
Wilmad Labglass FLSK VOL CL A 500ML ST19 STPR
Supplier:  Wilmad Labglass LG8110118
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 500mL +/- 0.20mL capacity. Stopper Size: 19.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A (Unserialized flasks) and ASTM E-288.
Catalog No. 50-806-490