Learn More
Wilmad Labglass FLSK VOL CL A 50ML PLS STPR
Supplier:  Wilmad Labglass LG8116110
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 50mL +/- 0.05mL capacity. Stopper Size: 9. Calibrated to contain at 20degreeC. Flask is provided with plastic closed-bottom Stopper..
Catalog No. 50-806-064
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product