Learn More
Wilmad Labglass FLSK VOL CL A SRLZD 1000ML
Supplier:  Wilmad Labglass LG8112120
Encompass
Volumetric Flask, Class A, To Contain, 1000mL +/- 0.30mL capacity. Stopper Size: 22. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.
Catalog No. 50-803-280