Learn More
Wilmad Labglass FLSK VOL CL A SRLZD 100ML
Supplier:  Wilmad Labglass LG8112112
Encompass
Volumetric Flask, Class A, To Contain, 100mL +/- 0.08mL capacity. Stopper Size: 13. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.
Catalog No. 50-803-171