Learn More
Wilmad Labglass FLSK VOL CL A SRLZD 2000ML
Supplier:  Wilmad Labglass LG8112122
Encompass
Volumetric Flask, Class A, To Contain, 2000mL +/- 0.50mL capacity. Stopper Size: 27. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.
Catalog No. 50-803-471
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.