Learn More
Wilmad Labglass FLSK VOL CL A SRLZD 200ML
Supplier:  Wilmad Labglass LG8112114
Encompass
Volumetric Flask, Class A, To Contain, 200mL +/- 0.10mL capacity. Stopper Size: 13. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.
Catalog No. 50-803-548