Learn More
Wilmad Labglass FLSK VOL CL A SRLZD 2ML
Supplier:  Wilmad Labglass LG8114102
Encompass
Volumetric Flask, Class A, To Contain, 2mL +/- 0.015mL capacity. Stopper Size: 8. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.
Catalog No. 50-803-072