Learn More
Wilmad Labglass FLSK VOL CL A SRLZD 50ML
Supplier:  Wilmad Labglass LG8112110
Encompass
Volumetric Flask, Class A, To Contain, 50mL +/- 0.05mL capacity. Stopper Size: 9. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.
Catalog No. 50-806-932