Learn More
Wilmad Labglass FLSK VOL CL B 100ML ST13 STPRR
Supplier:  Wilmad Labglass LG8125106
Encompass_Preferred
Volumetric Flask, Class B, To Contain, 100mL +/- 0.16mL capacity. Stopper Size: 13. Calibrated to contain at 20degreeC. Flask size complies with ASTM E-288. Supplied with one solid glass Pennyhead Stopper.
Catalog No. 50-807-689