Learn More
Wilmad Labglass FLSK VOL CL B 50ML ST9 STPR
Supplier:  Wilmad Labglass LG8125104
Encompass_Preferred
Volumetric Flask, Class B, To Contain, 50mL +/- 0.10mL capacity. Stopper Size: 9. Calibrated to contain at 20degreeC. Flask size complies with ASTM E-288. Supplied with one solid glass Pennyhead Stopper.
Catalog No. 50-806-933