Learn More
MSC Value Collection Polybag, 30 x 36 in, 3 mil, 100/Pack
Supplier:  MSC 39561972
Encompass
Polybags offer a versatile and flexible storage solution, protecting contents from dirt, dust, and moisture across diverse applications, from food to industrial tools and electronic equipment.
  • Manufactured from 100% virgin, high-clarity polyethylene film
  • Meets FDA and USDA specifications
  • Can be closed with twist ties, bag tape, or an impulse sealer
  • Recyclable material
  • Sturdy 3 mil thickness for medium to heavier products
  • Measured width x length, representing inside dimensions
Catalog No. 50-191-6653