Learn More
MSC Value Collection Polybag, 3 x 16 in, 3 mil, 1000/Pk.
Supplier:  MSC 39562343
Encompass
Polybags are versatile, flexible storage solutions suitable for various applications, from food to industrial tools, and electronic equipment. They are designed to protect contents from dirt, dust, and moisture, manufactured from 100% virgin, high clarity polyethylene film, and meet FDA and USDA specifications. The top of the bag is open and can be sealed using twist ties, bag tape, or an impulse sealer.
  • Suitable for food, industrial tools, and electronic equipment
  • Protects contents from dirt, dust, and moisture
  • Manufactured from 100% virgin, high clarity polyethylene film
  • Meets FDA and USDA specifications
  • Open top can be closed with twist ties, bag tape, or impulse sealer
  • Recyclable
Catalog No. 50-191-6690