Learn More
Medchemexpress LLC FLT3LG Human CHO 100ug
SDP
Supplier:  Medchemexpress LLC HYP7111100UG
Encompass_Preferred
Recombinant human FLT3 ligand expressed in CHO cells and supplied as a tag-free, lyophilized powder. The protein stimulates early hematopoietic progenitor proliferation via FLT3 activation, is formulated with trehalose for stability, and is characterized by high purity and potent bioactivity suitable for hematopoiesis and cytokine signaling research.
  • CHO-expressed, tag-free recombinant protein.
  • Lyophilized formulation with trehalose for enhanced stability.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Potent bioactivity: ED50 <1 ng/mL in cell proliferation assays.
  • Apparent molecular weight ~18-22 kDa due to glycosylation.
  • Store at -20°C unopened; aliquot and freeze after reconstitution for long-term storage.
Catalog No. 50-001-96665