Learn More
Chemglass Life Sciences 3L Filter Funnel, Buchner, Medium Frit
SDP
Supplier:  Chemglass Life Sciences CG140235
Encompass_Preferred
3L Filter Funnel, Buchner, Medium Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.
Catalog No. CG140235