Learn More
Cytiva (Formerly Pall Lab) Nylaflo™ Nylon Membrane Disc Filter - 0.2 m, 142 mm (25/pkg)
Supplier:  Cytiva 66604
Encompass
A Highly Versatile Laboratory Membrane Filter-Excellent chemical compatibility with esters, bases, and alcohols-Naturally hydrophilic-Available in 0.2 and 0.45 m pore sizes, in diameters ranging from 13 to 142 mm-HPLC certified. Provides assurance that the filter will not add artifacts to your analysis This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3345