Learn More
Apexbio Technology LLC Fluazinam 79622-59-6 500mg
SDP
Supplier:  Apexbio Technology LLC C6578500
Encompass_Preferred
Fluazinam (CAS 79622-59-6) is a small-molecule inhibitor targeting fungal mitochondrial respiratory chain complex II (succinate dehydrogenase SDH) It is designed to disrupt electron transfer during cellular respiration thereby impairing ATP production crucial for fungal cell viability Fluazinam exerts its biological activity primarily through inhibition of mitochondrial SDH In biochemical assays Fluazinam demonstrates inhibitory activity with reported IC50 values ranging from approximately 0 03 to 0 12 M against fungal SDH Based on these pharmacological properties Fluazinam holds research potential in investigating antifungal resistance mechanisms mitochondrial respiration impairment energy metabolism alterations and oxidative stress-related signal transduction pathways
Catalog No. 50-000-89704