Learn More
Medchemexpress LLC Fluazinam | 79622-59-6 | 99.4% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB183910MM/1ML
Encompass_Preferred
Fluazinam is an orally active dinitroaniline fungicide. It induces phosphorylation of JNK, activates the p38 pathway, decreases Bcl-2, activates caspase-3, decreases complex I activity, and increases Autophagy and Apoptosis. Fluazinam exhibits strong antifungal activity against F. fujikuroi and B. maydis. It also has a negative impact on Brachydanio rerio and worker bees.
  • Induces phosphorylation of JNK
  • Activates p38 pathway
  • Decreases Bcl-2
  • Activates caspase-3
  • Decreases complex I activity
  • Increases autophagy and apoptosis
  • Exhibits strong antifungal activity
Catalog No. 50-004-10798