Learn More
Medchemexpress LLC Fluazinam | 79622-59-6 | 99.4% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB18395G
Encompass_Preferred
Fluazinam is an orally active dinitroaniline fungicide. It induces phosphorylation of JNK, activates the p38 pathway, decreases Bcl-2, activates caspase-3, decreases complex I activity, and increases Autophagy and Apoptosis. Fluazinam exhibits strong antifungal activity against *F. fujikuroi* and *B. maydis*. It also has a negative impact on *Brachydanio rerio* and worker bees.
  • Induces SH-SY5Y cell death and nuclear damage.
  • Demonstrates great control efficiency against *B. maydis* on detached corn leaves.
  • Shows strong antifungal activity against *F. fujikuroi*.
  • Impairs oxidative phosphorylation and induces hyper/hypo-activity in zebrafish larvae.
  • Exhibits acute toxicity to *Brachydanio rerio*.
  • Reduces food consumption, damages midgut epithelium, increases autophagy and apoptosis in the midgut, induces oxidative stress, and alters walking behavior in worker bees.
Catalog No. 50-004-11758