Learn More
Selleck Chemical LLC Flubendazole S1837-50mg
SDP
Supplier:  Selleck Chemical LLC S183750MG
Encompass_Preferred
Flubendazole (Flumoxanal NSC 313680) is an autophagy inducer by targeting Atg4B used to treat internal parasite and worm infection
Catalog No. 50-310-9766