Learn More
Medchemexpress LLC Fluc-IN-1 | 2940242-10-2 | 98.4% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17049610MG
Encompass_Preferred
Fluc-IN-1 is a small-molecule inhibitor of firefly luciferase (IC50 25 nM) that also suppresses WNT/β-catenin signaling (IC50 33 nM in TOPFlash assay). It is formulated and characterized for use in biochemical and cell-based assays as a probe for luciferase-dependent reporter systems and WNT pathway biology.
  • Potent luciferase inhibition (IC50 25 nM).
  • Inhibits WNT/β-catenin signaling (IC50 33 nM, TOPFlash).
  • High purity suitable for biochemical and cell-based assays.
  • Supplied with structural and analytical data for verification.
  • Available in multiple pack formats, including solid and DMSO solution.
Catalog No. 50-002-95386