Learn More
Selleck Chemical LLC Fluconazole S1331-50mg
SDP
Supplier:  Selleck Chemical LLC S133150MG
Encompass_Preferred
Fluconazole is a fungal lanosterol 14 alpha-demethylase inhibitor which thereby prevents the formation of ergosterol used in the treatment and prevention of superficial and systemic fungal infections
Catalog No. 50-311-1271