Learn More
Cayman Chemical Fluconazole-d4 5mg
SDP
Supplier:  Cayman Chemical 182525 MG
Encompass_Preferred
An internal standard for the quantification of fluconazole by GC- or LC-MS
Catalog No. 50-296-8368