Learn More
Apexbio Technology LLC Fluconazole hydrate 155347-36-7 100mg
SDP
Supplier:  Apexbio Technology LLC B1226100
Encompass_Preferred
Fluconazole hydrate (CAS 155347-36-7) is a small-molecule inhibitor targeting the fungal enzyme lanosterol 14 -demethylase It is designed to inhibit ergosterol biosynthesis thereby disrupting fungal cell membrane integrity and arresting fungal growth Fluconazole hydrate exerts its biological activity primarily through inhibition of lanosterol 14 -demethylase In in vitro studies fluconazole hydrate demonstrates inhibitory activity with documented IC50 values typically ranging from 0 1 to 10 g/mL depending on fungal strain and experimental conditions Based on these pharmacological properties fluconazole hydrate holds research potential in antifungal pharmacology assays fungal infection model studies and evaluation of combinational antifungal therapies
Catalog No. 50-000-92166