Learn More
Medchemexpress LLC Fludarabine | 21679-14-1 | 99.7% | C10H12FN5O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0069
Encompass_Preferred
Fludarabine is a DNA synthesis inhibitor and a fluorinated purine analogue, exhibiting antineoplastic activity in lymphoproliferative malignancies. It inhibits cytokine-induced activation of STAT1 and STAT1-dependent gene transcription in normal resting or activated lymphocytes.
  • Acts as a DNA synthesis inhibitor
  • Functions as a fluorinated purine analogue
  • Exhibits antineoplastic activity in lymphoproliferative malignancies
  • Inhibits cytokine-induced activation of STAT1 and STAT1-dependent gene transcription
  • Induces a decrease in p27kip1 expression in vitro
  • Decreases incidence of graft-versus-host disease in animal models
Catalog No. 50-003-58002