Learn More
Medchemexpress LLC Fludarabine chloride | 2734853-80-4 | 99.7% | 303.68 g/mol | C10H11ClFN5O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1454435MG
Encompass_Preferred
Fludarabine chloride is a research-grade nucleoside analogue used in biochemical and cellular studies. It inhibits RNA adenosine deaminase 1 (ADAR1) with reported in vitro activity and is supplied in solid and solution formats for laboratory research. Typical applications include RNA-editing assays, cancer-related cellular studies, and inhibitor screening.
  • Inhibits ADAR1 with reported IC50 ≈ 0.87 μM.
  • Available as solid and as a 10 mM solution for flexible use.
  • High reported purity suitable for research applications.
  • Supplied in small research quantities for assay development.
  • Suitable for cellular assays and biochemical inhibition studies.
Catalog No. 50-002-21580