Learn More
Medchemexpress LLC Fludrocortisone | 127-31-1 | 99.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1203
Encompass_Preferred
Fludrocortisone is an orally active mineralocorticoid and glucocorticoid receptor agonist. It is used in research for conditions such as congenital adrenal hyperplasia, postural hypotension, and adrenal insufficiency. It works by:
  • Suppressing pro-inflammatory cytokine expression and reducing CCL2, IL-6, IL-8 levels.
  • Upregulating mineralocorticoid receptor (MR) expression.
  • Inducing PI3K/Akt, GSK-3β, CREB, ERK1/2, mTOR phosphorylation.
  • Blocking Tau hyperphosphorylation, preventing apoptosis, and promoting survival and proliferation.
  • Enhancing renal sodium and water transport, increasing plasma volume and blood pressure, and reducing plasma potassium and renin activity.
  • Stimulating erythropoietin expression.
  • Modulating uterine receptivity genes and reversing MUC1 upregulation.
Catalog No. 50-004-14297