Learn More
Selleck Chemical LLC Fludrocortisone acetate S3659-25mg
SDP
Supplier:  Selleck Chemical LLC S365925MG
Encompass_Preferred
Fludrocortisone (9 -fluorocortisol) is a synthetic pregnane steroid and a halogenated derivative of cortisol It is used to treat adrenogenital syndrome postural hypotension and adrenal insufficiency
Catalog No. 50-310-9729