Learn More
Fluotics Fluent Evo LiHa P1000 clear sterile
Supplier:  Fluotics LHA1000.ST
Encompass
Tecan Fluent Evo LiHa P1000, clear, sterile, Hanging Rack, for FCA/LiHa, working volume: 1000 uL, Case Size: 24 racks/case with 96 tips per rack, Compare to: 30126094
Catalog No. NC2195939