Learn More
Fluotics Fluent Evo LiHa P200 conductive sterile
Supplier:  Fluotics LHA200CBR.ST
Encompass
Tecan Fluent Evo LiHa P200, conductive, sterile, Single Stack, refill tray, for FCA/LiHa, working volume: 200 uL, Case Size: 24 racks/case with 96 tips per rack, Compare to: 30057814
Catalog No. NC2195965