Learn More
Fluotics Fluent Evo LiHa P50 clear filter/sterile
Supplier:  Fluotics LHA50R.F
Encompass
Tecan Fluent Evo LiHa P50, clear, filter/sterile, Single Stack, refill tray, for FCA/LiHa, working volume: 50 uL, Case Size: 24 racks/case with 96 tips per rack
Catalog No. NC2195984