Learn More
Cayman Chemical FlufenamIc AcId-d4 1mg
SDP
Supplier:  Cayman Chemical 332721 MG
Encompass_Preferred
An internal standard for the quantification of flufenamic acid by GC- or LC-MS
Catalog No. 50-294-7588