Learn More
Glass Expansion Fluka RBS-25 Concentrate 1Litre
SDP
Supplier:  Glass Expansion FLUKA25
Encompass_Preferred
Fluka RBS-25 Concentrate 1 Liter. A concentrated cleaning solution that is highly effective in removing clogs from nebulizers, and deposits/residues from spray chambers, torches, injectors, and cones.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC9000117
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.