Learn More
TARGETMOL CHEMICALS INC Flumazenil 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1240500MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Flumazenil (Ro 15-1788) antagonizes the benzodiazepine binding site of the gamma-aminobutyric acid (GABA)/benzodiazepine receptor complex in the central nervous system (CNS), thereby preventing the chloride channel opening events and inhibiting neuronal hyperpolarization. As a result, flumazenil reverses benzodiazepine-induced effects including sedation, psychomotor deficits, amnesia, and hypoventilation in a dose-dependent manner. Flumazenil is an imidazobenzodiazepine derivative, effective in reversing benzodiazepine-induced activities. Purity 99.62%
Catalog No. 50-287-8459