Learn More
Medchemexpress LLC Flumazenil acid | 84378-44-9 | MFCD08061420 | 100.0% | 275.24 | C13H10FN3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11884410MG
Encompass_Preferred
Flumazenil acid is the acidic metabolite of flumazenil and is supplied as a solid research chemical for pharmacology, metabolism, and analytical studies.
  • High purity suitable for analytical and pharmacological assays.
  • Characterized by molecular formula C13H10FN3O3 and molecular weight 275.24 for reference and identification.
  • Available in small, research-scale quantities for in vitro and in vivo studies.
  • Provided as a stable solid with consistent lot-specific certification and quality data.
  • Delivered with supporting documents such as a data sheet, certificate of analysis, and safety data sheet for compliant laboratory use.
Catalog No. 50-002-29209