Learn More
Medchemexpress LLC Flumazenil acid | 84378-44-9 | MFCD08061420 | 100.0% | 275.24 g/mol | C13H10FN3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11884450MG
Encompass_Preferred
Flumazenil acid is the primary metabolite of flumazenil and is supplied as a solid research reference standard for analytical and pharmacological studies. It is intended for use in metabolite profiling, impurity characterization, and method development.
  • Primary metabolite of flumazenil.
  • Used as a reference standard for analytical and metabolic studies.
  • High purity suitable for research (99.96%).
  • Molecular weight 275.24 g/mol; formula C13H10FN3O3.
  • Off-white to light yellow solid with recommended cold storage for stability.
  • Documentation available for analytical use, including data sheet, COA, and SDS.
Catalog No. 50-003-08069