Learn More
Cayman Chemical FlumazenIl-d5 2.5mg
SDP
Supplier:  Cayman Chemical 285292.5 MG
Encompass_Preferred
An internal standard for the quantification of flumazenil by GC- or LC-MS
Catalog No. 50-294-0279