Learn More
Selleck Chemical LLC Flumazenil 100mg 78755-81-4 RO 15-1788
SDP
Supplier:  Selleck Chemical LLC S1332100MG
Encompass_Preferred
Flumazenil (RO 15-1788) is a competitive GABAA receptor antagonist, used in the treatment of benzodiazepine overdoses. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-764-9