Learn More
Medchemexpress LLC HY-B0526 100mg Medchemexpress, Flumequine CAS:42835-25-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0526100MG
Encompass_Preferred
Medchemexpress, HY-B0526 100mg Flumequine CAS:42835-25-6 Flumequine (R-802) is a quinolone antibiotic, and acts as a topoisomerase II inhibitor, with an IC50 of 15 μM (3.92 μg/mL). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2580