Learn More
Medchemexpress LLC Flumioxazin | 25MG
SDP
Supplier:  Medchemexpress LLC HY11450725MG
Encompass_Preferred
Flumioxazin (Sumisoya) is an herbicide for use in soybean and peanut Flumioxazin inhibits the enzyme protoporphyrinogen oxidase[1][2] Flumioxazin is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-30003