Learn More
Selleck Chemical LLC Flunarizine 2HCl 25mg 30484-77-6 KW-3149, R14950
SDP
Supplier:  Selleck Chemical LLC S203025MG
Encompass_Preferred
Flunarizine 2HCl (KW-3149, R14950) is a dihydrochloride salt form which is a calcium channel blocker with a Ki of 68 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8210