Learn More
Apexbio Technology LLC Azelnidipine 123524-52-7 10mg
SDP
Supplier:  Apexbio Technology LLC B141410
Encompass_Preferred
Azelnidipine (CAS 123524-52-7) is a dihydropyridine-based small molecule that functions as an L-type calcium channel blocker By inhibiting calcium influx through these channels azelnidipine effectively modulates vascular smooth muscle tone resulting in vasodilation and reduced blood pressure In preclinical studies acute administration of azelnidipine has been shown to prevent abrupt declines in cardiac function following acute stress events This compound is utilized in biomedical research to investigate the cellular and systemic mechanisms underlying calcium channel modulation and the physiological response to antihypertensive therapies
Catalog No. 50-000-92320