Learn More
Apexbio Technology LLC Flunixin Meglumin, 1g Cas# 42461-84-7
SDP
Supplier:  Apexbio Technology LLC B14451000
Encompass_Preferred
Flunixin Meglumine is a potent inhibitor of COX used as analgesic agent with anti-inflammatory and antipyretic activity. 42461-84-7. MFCD01725419 IC50: A potent cyclooxygenase inhibitor with IC50 values of 1 nM.Flunixin meglumine serves as a non-narcotic and non-steroidal analgesic agent with antipyretic activities. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8615