Learn More
Medchemexpress LLC Flunixin meglumine | 42461-84-7 | 99.5% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0386R100MG
Encompass_Preferred
Flunixin meglumine (Standard) is an analytical standard and a cyclooxygenase (COX) inhibitor. It demonstrates anti-inflammatory effects, making it suitable for various research and analytical applications.
  • Analytical standard for research
  • Acts as a cyclooxygenase (COX) inhibitor
  • Exhibits anti-inflammatory effects
  • Used as a reference standard in HPLC, GC, and MS experiments
Catalog No. 50-004-06426